Centrifuge tubes, graduated, Goetz type, KIMBLE®
Supplier: KIMBLEKONTES
45220-100
KIML45220-100EA 1480 SGD
KIML45220-100
Centrifuge tubes, graduated, Goetz type, KIMBLE®
Tubes Centrifuge Tubes
Borosilicate glass, KIMAX®, graduated.
  • Calibrated to contain
  • Durable black ceramic enamel scale
Used for the determination of small quantities of solids in large volumes of liquids. It is recommended for the determination of free water and sediment in diesel and other distillate fuels, as a pass-fail indication of product quality.
Do not centrifuge with stopper.
Certifications: Glass conforms to USP Type I and ASTM E438, Type I, Class A requirements.