Centrifuge tube, oil, graduated, KIMBLE®
Supplier: KIMBLEKONTES
45240-100
KIML45240-100EA 1580 SGD
KIML45240-100
Centrifuge tube, oil, graduated, KIMBLE®
Tubes Centrifuge Tubes
Borosilicate glass KIMAX®, graduated, snap cap finish.
  • Scale and legend of durable white ceramic enamel
  • Top is tooled to accept a snap cap
Used in the determination of water and sediment in crude mineral oils, fuel oils and other petroleum products (D1796 and MPMS 10.4 standards)
Used in determination of volume of precipitate formed by centrifuging definite quantities of steam cylinder stocks and black oils and other lubricating oils (ASTM D91 and D128)
Used in testing for acidity of distillation residues or hydrocarbon liquids of gasoline or petroleumMore Product Information