Centrifuge tube, oil and weathering (end point index), KIMAX®, KIMBLE®
Supplier: KIMBLEKONTES
45241-100
KIML45241-100EA 1540 SGD
KIML45241-100
Centrifuge tube, oil and weathering (end point index), KIMAX®, KIMBLE®
Tubes Centrifuge Tubes
Borosilicate glass KIMAX®, graduated, snap cap finish.
  • Scale and legend of durable white ceramic enamel
  • Top is tooled to accept a snap cap
For use in the determination of residues in liquified petroleum (LP) gases, ASTM Method D2158
Dimensions (O.D.×L): 37×203 mm