Nessler colour comparison tubes, KIMBLE®
Supplier: DWK Life Sciences
SP45310-50100 45310-100 45310-50
SCERSP45310-50100EA 0 SGD
SCERSP45310-50100 KIML45310-100 KIML45310-50
Nessler colour comparison tubes, KIMBLE®
Tubes Nessler Tubes
Manufactured from 33 expansion, low extractable borosilicate glass.
  • Shadowless bottom of each tube transmits undistorted light
  • Rings and legend are durable white ceramic enamel
  • Matched or unmatched available
These Nessler tubes are suitable for colour comparison.
Certifications: Glass conforms to USP Type I and ASTM E438 Type I Class A requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable