Nessler tubes, KIMBLE®
Supplier: KIMBLEKONTES
45315-100 45315-50
KIML45315-100EA 478 SGD
KIML45315-100 KIML45315-50
Nessler tubes, KIMBLE®
Tubes Nessler Tubes
These Nessler tubes are ideal for colour comparison. Graduated ring scale of any of the tubes within a set will not vary from that of any other tube within the set by more than 1,5 mm for the 50 ml and 2,0 mm for the 100 ml sizes.
  • Shadowless bottom of tubes transmits undistorted light
  • Rings and markings are in durable white ceramic enamel which correspond with specific volumes of liquid
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I Class A requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable