Plain high strength centrifuge tubes
Supplier: KIMBLEKONTES
45500-15
KIML45500-15EA 160 SGD
KIML45500-15
Plain high strength centrifuge tubes
Tubes Centrifuge Tubes
Plain high strength centrifuge tubes can be centrifuged up to 13100 RCF when used with an accessory rubber adapter sleeve in a 50 ml rotor cavity.
  • Without closures
  • Tubes can withstand temperatures up to 300 °C
Tubes have been chemically strengthened to achieve a greater mechanical strength than standard borosilicate centrifuge tubes.
Reusable centrifuge tubes are manufactured from ASTM E438 Type I, Class B, borosilicate glass.
Order Now
SPECIFICATIONS
Environmentally Preferable