Centrifuge tubes, round-bottom, KIMBLE®
Supplier: KIMBLEKONTES
45600-30
KIML45600-30EA 385 SGD
KIML45600-30
Centrifuge tubes, round-bottom, KIMBLE®
Tubes Centrifuge Tubes
These tubes are manufactured from chemically strengthened borosilicate glass.
  • Reusable
  • Can be centrifuged up to 13100×g when used with the accessory rubber adapter sleeve in a 50 ml rotor cavity
  • Withstand temperatures up to 300 °C
  • Screw-thread tubes are supplied with PTFE-faced rubber liner caps
Chemically strengthened glass to achieve a greater mechanical strength than standard borosilicate centrifuge tubes.
Glass conforms to ASTM E438 Type I, Class B requirements.
Ordering information: Accessories sleeves and replacement caps sold separately.