Bottles, narrow neck, round, Boston type, KIMBLE®
Supplier: KIMBLEKONTES
5110120V-21 5110220V-21 5110422V-21 5110824V-21 5111628V-21 5113233V-21 5120120V-21 5120220V-21 5120422V-21 5120824V-21 5121628V-21 5123233V-21
KIML5110120V-21EA 108 SGD
KIML5110120V-21 KIML5110220V-21 KIML5110422V-21 KIML5110824V-21 KIML5111628V-21 KIML5113233V-21 KIML5120120V-21 KIML5120220V-21 KIML5120422V-21 KIML5120824V-21 KIML5121628V-21 KIML5123233V-21
Bottles, narrow neck, round, Boston type, KIMBLE®
Bottles Glass Bottles
Soda-lime glass, Type-III, clear or amber.
  • Packaged in convenience packs with poly vinyl caps attached
  • Helps to eliminate waste and insures product integrity for long term storage
  • Narrow-mouth design for easy pouring
Clear bottles allow for viewing of contents while amber bottles are designed to protect contents from UV rays and are ideal for light-sensitive products.
Order Now
SPECIFICATIONS
Environmentally Preferable