Bottles, wide neck, clear, round, graduated, AC medium, KIMBLE®
Supplier: KIMBLEKONTES
5810133V-26 5810238V-26 5810448V-26 5810858V-26 5811670V-26
KIML5810133V-26EA 299 SGD
KIML5810133V-26 KIML5810238V-26 KIML5810448V-26 KIML5810858V-26 KIML5811670V-26
Bottles, wide neck, clear, round, graduated, AC medium, KIMBLE®
Bottles Glass Bottles
Soda-lime glass.
  • PTFE faced LDPE foam closure attached
  • Clear glass allows for easy viewing of contents
  • Graduated in both milliliters and ounces
  • Available as convenience packs
Ideal for storage for solids, liquids, powders and specimens. It features wide-mouth design for efficient addition and removal of contents.
Order Now
SPECIFICATIONS
Environmentally Preferable