Bottles, narrow neck, KIMBLE®
Supplier: KIMBLEKONTES
5916438B
KIML5916438BEA 83.1 SGD
KIML5916438B
Bottles, narrow neck, KIMBLE®
Bottles Glass Bottles
Soda-lime glass, clear or amber.
  • Narrow mouth design for safe pouring
  • Good chemical and physical properties
  • For applications that do not involve high thermal stresses
Clear bottles allow for viewing of contents while amber bottles are designed to protect contents from UV rays and are ideal for light-sensitive products.