Culture tubes, disposable, prelabeled, Mark-M™, KIMBLE®
Supplier: KIMBLEKONTES
60AM10MXW
KIML60AM10MXWEA 229 SGD
KIML60AM10MXW
Culture tubes, disposable, prelabeled, Mark-M™, KIMBLE®
Tubes Culture Tubes
Manufactured from 90 expansion soda-lime glass.
  • Permanent marking vertical spot
  • Sturdy, uniform bottoms and consistent lengths
Certifications: Glass conforming USP Type III requirements.