Pasteur pipettes, disposable, KIMBLE®
Supplier: KIMBLEKONTES
63B93P
KIML63B93PEA 474 SGD
KIML63B93P
Pasteur pipettes, disposable, KIMBLE®
Pipettes Pasteur/Transfer Pipettes
Soda Lime glass, disposable.
  • Intended for one-time use in the transfer of small volumes
  • With or without cotton plug
  • Soda lime pipettes are manufactured from 90 expansion soda lime glass conforming to USP Type III requirements
Order Now
SPECIFICATIONS
Environmentally Preferable