Culture tubes, disposable, KIMBLE®
Supplier: KIMBLEKONTES
73500-18150 73500-1075 73500-16125 73500-16150 73500-1275 73500-16100 73500-650 60D47729-574 73500-20150
KIML73500-18150EA 302 SGD
KIML73500-18150 KIML73500-1075 KIML73500-16125 KIML73500-16150 KIML73500-1275 KIML73500-16100 KIML73500-650 KIML60D47729-574 KIML73500-20150
Culture tubes, disposable, KIMBLE®
Tubes Culture Tubes
Manufactured from 51 expansion borosilicate glass.
  • Offers excellent chemical resistance against sodium leaching, a factor common in soda lime glass products
  • Tubes are packed in convenient trays for ease of use on bench tops and in drawers
  • Trays are shrink-wrapped in plastic film, with tubes remaining in constant alignment for protection against in-transit breakage and contamination
  • The 6×50 mm size is often referred to as a 'Durham tube'
These premium quality tubes feature sturdy, uniform bottoms and consistent lengths. The 10×75 and 12×75 mm sizesMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable