Tissue Grinders with PTFE Pestles, KONTES®, DUALL®, KIMBLE®
Supplier: KIMBLEKONTES
885480-0022 885480-0023 885480-0020 885480-0021
KIML885480-0022EA 425 SGD
KIML885480-0022 KIML885480-0023 KIML885480-0020 KIML885480-0021
Tissue Grinders with PTFE Pestles, KONTES®, DUALL®, KIMBLE®
Homogenisers Tissue Grinders
Tissue grinders greatly improve grinding efficiency by combining both conical and cylindrical surfaces to effectively reduce tissue and produce a uniform homogenate.
  • PTFE pestle is ideal for soft tissue such as brain or liver
  • Construction is strong enough to allow the pestle to be motor driven
Clearance between pestle and tube is 0,1 to 0,15 mm.
Ordering information: Special clearance pestles are available upon request; contact your Avantor sales representative for more information.
Order Now
SPECIFICATIONS
Environmentally Preferable