Filtering flasks, ULTRA-WARE®, KimCote®, KIMBLE®
Supplier: KIMBLEKONTES
953760-1002
KIML953760-1002EA 492 SGD
KIML953760-1002
Filtering flasks, ULTRA-WARE®, KimCote®, KIMBLE®
Flasks Filter Flasks
Manufactured from 33 expansion, low extractable borosilicate glass.
  • Excellent chemical resistance
  • Excellent temperature resistance
These erlenmeyer flasks are heavy walled to ensure the mechanical strength needed for vacuum filtration and the safety coating reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals. All flasks have a side arm for connection to 6 mm int.Ø flexible tubing.
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable