Archiving and storage KP systems: filing cabinets
Supplier: KLINIPATH
KLIN10-0017EA 11180 SGD
KLIN10-0017 KLIN10-0019
Archiving and storage KP systems: filing cabinets
Furniture Storage Cabinets
Every laboratory has its specific needs and demands to handle slide and cassette storage. For high volume storage of slides and/or cassettes we offer various types of cabinets with drawers and inserts adapted to specific needs.
Complete cabinets for cassettes and slides, including cassette boxes or slide inserts.
  • Robust design, high quality finishing
  • Drawer safety lock to prevent from opening more than one drawer at the same time
  • Drawers run on ball bearings (makes it slide smoothly)
  • Drawer fronts are available in blue, purple-red, dark grey, green, light
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable