KP-Mega moulds
Supplier: KLINIPATH
3055
KLIN3055EA 101 SGD
KLIN3055
KP-Mega moulds
Moulds Moulds for Histology
These moulds are made of stainless steel and are suitable for Mega cassettes.
Order Now
SPECIFICATIONS
Environmentally Preferable