Microscope slide with positive charge
Supplier: KLINIPATH
KLINPR-P-001EA 39.7 SGD
KLINPR-P-001
Microscope slide with positive charge
Slides Adhesive Slides
This microscope slide is designed with grounded-90, finished and clipped corners of 1 mm.
  • Suitable for use in all types of slide printers
  • Used for IHC staining and other special techniques
  • Easy handling in slide racks, automated slide printers and digital slide imagers
Packaging: In sealed plastic dispenser boxes.
Order Now
SPECIFICATIONS
Environmentally Preferable