Silane adhesive slides, KF Frost
Supplier: KLINIPATH
KLINPR-S-001EA 512 SGD
KLINPR-S-001
Silane adhesive slides, KF Frost
Slides Adhesive Slides
These silane adhesive slides are specially designed for thermal transfer printers.
Certifications: CE certified according to IVDR (2017/746).