Counting chambers
Supplier: Kova International
87144E 87146E 87100E 87118E 87157E
KOVI87144EEA 373 SGD
KOVI87144E KOVI87146E KOVI87100E KOVI87118E KOVI87157E
Counting chambers
Slides Counting Chambers
Designed to reduces pipetting errors and eliminate variable that could interfere with microscopic urinalysis (UA).
  • High optical quality through the composition acrylate
  • Perimeter contained wells for precise volume control and protection of cross-contamination and leakage
  • Capillary action for reproducible precision
  • Homogeneous suspension of sample sediments: Retained volume of 1 µl
  • For grid models, the grid is made up of nine squares, each divided into nine smaller squares
Grid: 3×3 mm
Volume retained on the grid: 1 µl