Counting chamber, Fast Read® 102
Supplier: Kova International
BVS100H
KOVIBVS100HEA 229 SGD
KOVIBVS100H
Counting chamber, Fast Read® 102
Slides Counting Chambers
Fast Read 102 systems are made of a disposable plastic slide and designed for the standardisation of microscopic urinalysis.
  • Protected by an optically transparent film
  • Precision and reproducibility guarantee constant readings
  • Identification of the sample by using the numbers printed on the sides
Each chamber is fitted with its own system for the collection of excess urine to prevent any possible contamination.
Chamber depth: 0,1 mm
Chamber volume: 7 µl
Order Now
SPECIFICATIONS
Environmentally Preferable