Water baths, insulated stainless steel
Supplier: LAUDA
LCZ0712
LAUDLCZ0712EA 0 SGD
LAUDLCZ0712
Water baths, insulated stainless steel
Baths Water Baths, Unstirred
Insulated stainless steel baths which can be used up to 200 °C. The outer jacket is made of powder coated sheet steel. All baths are equipped with a built-in drain tap. These high quality, robust baths are available in a variety of sizes and can be combined with immersion thermostats from Lauda or other manufacturers.