Centrifuge tubes, SuperClear®
Supplier: LABCON
3192-870-008-9
LBCN3192-870-008-9EA 578 SGD
LBCN3192-870-008-9
Centrifuge tubes, SuperClear®
Tubes Centrifuge Tubes
SuperClear® high performance centrifuge tubes are lot-tested at a much higher g-force than rated to ensure that they can be spun safely. The quality control team tests the seals with chloroform. 
  • ATP-, RNase/DNase- and endotoxin-free (non-pyrogenic)
  • Patented anti-smear writing surface
  • 3 colour printed graduations and reverse reading scale
  • Convenient freeze line
  • Ultra-clear Polypropylene USP Class VI resin, free of DiHEMDA, oleamide, and BPA
  • Freezable to –80 °C, and autoclavable
SuperClear® tubes are made from an ultra-clear, medical grade resinMore Product Information