Glass stoppers, standard taper, hexagonal, hollow
Supplier: Lenz Laborglas GmbH & CO.KG
01400007 01400014 01400345 01400307 01400324 01400314 01400312 01400329 01400310 01400319 01400334
LENZ01400007EA 8.1 SGD
LENZ01400007 LENZ01400014 LENZ01400345 LENZ01400307 LENZ01400324 LENZ01400314 LENZ01400312 LENZ01400329 LENZ01400310 LENZ01400319 LENZ01400334
Glass stoppers, standard taper, hexagonal, hollow
Stoppers Glass Stoppers
These standard taper hollow stoppers are made from borosilicate glass 3.3 and have a hexagonal head.
  • Excellent chemical resistance
  • High thermal resistance, minimal heat expansion results in high resistance to temperature changes
  • Hexagonal head allows easy release and removal of the stopper
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus.
Certifications: DIN 12252
Ordering information: Available with flat or pointed bottom.