Dropping funnels, cylindrical, without pressure balance
Supplier: Lenz Laborglas GmbH & CO.KG
04172814 04174914 04175814 04177014 04172811 04173711 04173714 04172211
LENZ04172814EA 0 SGD
LENZ04172814 LENZ04174914 LENZ04175814 LENZ04177014 LENZ04172811 LENZ04173711 LENZ04173714 LENZ04172211
Dropping funnels, cylindrical, without pressure balance
Funnels Dropping Funnels
DURAN®, borosilicate glass, clear, with PTFE key.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Cylindrical shape. With standard ground joint cone and socket. Clearly visible white divisions for approximate volume reading.
DIN ISO 4800
Certifications: DIN ISO 4800
Order Now
SPECIFICATIONS
Environmentally Preferable