Liebig condenser with standard ground joints
Supplier: Lenz Laborglas GmbH & CO.KG
05210202 05210301 05210303 05210304 05210402 05210403 05210404 05210504 05210401 05210704
LENZ05210202EA 0 SGD
LENZ05210202 LENZ05210301 LENZ05210303 LENZ05210304 LENZ05210402 LENZ05210403 LENZ05210404 LENZ05210504 LENZ05210401 LENZ05210704
Liebig condenser with standard ground joints
Condensers
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours, standard ground joint cone and sleeve. Water connection via glass olive.
Certifications: DIN 12576