Allihn condensers with standard ground joints
Supplier: Lenz Laborglas GmbH & CO.KG
05223302 05223303 05223304 05223403 05223404 05223504 05223201 05223202 05223204 05223301 05223203
LENZ05223302EA 0 SGD
LENZ05223302 LENZ05223303 LENZ05223304 LENZ05223403 LENZ05223404 LENZ05223504 LENZ05223201 LENZ05223202 LENZ05223204 LENZ05223301 LENZ05223203
Allihn condensers with standard ground joints
Condensers
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With five 'balls' (up to 250 mm jacket length) or eight 'balls' (for 400 mm jacket length), as well as standard ground joint cone and sleeve. Water connection via glass olive.
Compliant with DIN 12581 or supplementary DIN
Certifications: Compliant with DIN 12581 or supplementary DIN