Condensers, Allihn, for Soxleth extractors
Supplier: Lenz Laborglas GmbH & CO.KG
05226004 05226005 05226006
LENZ05226004EA 0 SGD
LENZ05226004 LENZ05226005 LENZ05226006
Condensers, Allihn, for Soxleth extractors
Condensers
DURAN® borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With NS cone. Water connection with glass nozzles.