Condensers, Allihn, for Soxleth extractors
Supplier: Lenz Laborglas GmbH & CO.KG
05226016 05226015 05226014
LENZ05226016EA 0 SGD
LENZ05226016 LENZ05226015 LENZ05226014
Condensers, Allihn, for Soxleth extractors
Condensers
DURAN® borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With NS cone. Water connection with two threaded GL 14 plastic olives.