Dimroth condensers with standard ground joints
Supplier: Lenz Laborglas GmbH & CO.KG
05240201 05240302 05240303 05240304 05240404 05240202 05240301 05240406 05240306
LENZ05240201EA 0 SGD
LENZ05240201 LENZ05240302 LENZ05240303 LENZ05240304 LENZ05240404 LENZ05240202 LENZ05240301 LENZ05240406 LENZ05240306
Dimroth condensers with standard ground joints
Condensers
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For cooling condensate from vapours, with core and jacket. Water supplied through glass connectors with 8 mm external.
Order Now
SPECIFICATIONS
Environmentally Preferable