Condensers, high efficient, four coil
Supplier: Lenz Laborglas GmbH & CO.KG
05255000 05255050
LENZ05255000EA 0 SGD
LENZ05255000 LENZ05255050
Condensers, high efficient, four coil
Condensers
DURAN® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
High-efficient condensers with four cooling coils, cooling surface: 2,350 cm².