Drying tubes, straight
Supplier: Lenz Laborglas GmbH & CO.KG
05330013
LENZ05330013EA 0 SGD
LENZ05330013
Drying tubes, straight
Tubes Drying Tubes
DURAN®, borosilicate glass, clear. One bulb and glass tubing connection.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes