Drying tubes with standard ground joints, U-shaped
Supplier: Lenz Laborglas GmbH & CO.KG
05342013 05342015
LENZ05342013EA 0 SGD
LENZ05342013 LENZ05342015
Drying tubes with standard ground joints, U-shaped
Tubes Drying Tubes
DURAN®, borosilicate glass, clear. Replaceable NS stopcocks. Lateral tube connectors with 6 mm external diameter.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Order Now
SPECIFICATIONS
Environmentally Preferable