Condensers with receivers, Twisselmann
Supplier: Lenz Laborglas GmbH & CO.KG
05420104 05420105 05420106
LENZ05420104EA 0 SGD
LENZ05420104 LENZ05420105 LENZ05420106
Condensers with receivers, Twisselmann
Condensers
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes