Extractor heads for specific light solvents
Supplier: Lenz Laborglas GmbH & CO.KG
05432137 05432149 05432158
LENZ05432137EA 0 SGD
LENZ05432137 LENZ05432149 LENZ05432158
Extractor heads for specific light solvents
Extractors Liquid-Liquid Extractors
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For the extraction of liquids with specific light solvents, with insert filter (porosity: 0).
Order Now
SPECIFICATIONS
Environmentally Preferable