Extractors for specific heavy solvents
Supplier: Lenz Laborglas GmbH & CO.KG
05433037 05433049 05433058
LENZ05433037EA 0 SGD
LENZ05433037 LENZ05433049 LENZ05433058
Extractors for specific heavy solvents
Extractors Liquid-Liquid Extractors
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For the extraction of liquids with specific heavy solvents.