Wash bottles, glass
Supplier: Lenz Laborglas GmbH & CO.KG
05475070
LENZ05475070EA 0 SGD
LENZ05475070
Wash bottles, glass
Bottles Wash Bottles
DURAN®, borosilicate glass clear.
  • Very good chemical resistance
  • Very good thermal resistance
  • Fully interchangeable parts
Universal wash bottle consisting of a 1000 ml capacity flat bottomed flask with standard ground joint sleeve NS 29/32 and spray assembly with standard ground joint cone NS 29/32.