Water jet pumps acc. to Friedrichs-Antlinger
Supplier: Lenz Laborglas GmbH & CO.KG
05580003
LENZ05580003EA 0 SGD
LENZ05580003
Water jet pumps acc. to Friedrichs-Antlinger
Pumps Vacuum Water Jet Pumps
Duran®, with hose connections. With spiral nozzle. For high performance.
  • Water consumption: 10 L/min at 4 bar water pressure
  • Suction speed: 400 ml/sec. at 4 bar water pressure
  • Final pressure 16 mbar at 4 bar water pressure