Dissolved oxygen bottles
Supplier: Lenz Laborglas GmbH & CO.KG
05605049 05605037
LENZ05605049EA 0 SGD
LENZ05605049 LENZ05605037
Dissolved oxygen bottles
Bottles Dissolved Oxygen Bottles
Glass, clear.
  • White inscription label
  • Oblique ground NS stopper
  • Optional stopper clamp is available
For the determination of dissolved oxygen in water according to Winkler. The capacity is engraved on the bottle with an accuracy of ± 0,01 ml. Each bottle is individually adjusted with its stopper (non interchangeable). Each stopper and bottle is identified with the same serial number.
Order Now
SPECIFICATIONS
Environmentally Preferable