KPG stirrers with lateral blade
Supplier: Lenz Laborglas GmbH & CO.KG
06307201 06307010 06307301 06307303 06307305 06307311 06307001 06307005 06307008 06307011 06307013 06307015 06307205 06307208 06307210 06307211 06307213 06307215 06307310 06307400
LENZ06307201EA 0 SGD
LENZ06307201 LENZ06307010 LENZ06307301 LENZ06307303 LENZ06307305 LENZ06307311 LENZ06307001 LENZ06307005 LENZ06307008 LENZ06307011 LENZ06307013 LENZ06307015 LENZ06307205 LENZ06307208 LENZ06307210 LENZ06307211 LENZ06307213 LENZ06307215 LENZ06307310 LENZ06307400
KPG stirrers with lateral blade
Stirrers Stirring Tools
DURAN®, borosilicate glass with PTFE paddle and paddle safety securing screw. 10 mm shaft Ø. The length of the ground and polished bearing surface for the shaft Ø is 160 mm.
Order Now
SPECIFICATIONS
Environmentally Preferable