Stirrers with lateral blade and GL thread
Supplier: Lenz Laborglas GmbH & CO.KG
06307061 06307063
LENZ06307061EA 0 SGD
LENZ06307061 LENZ06307063
Stirrers with lateral blade and GL thread
Stirrers Stirring Tools
Stirrers made of precision calibrated glass tube, length of the ground and polished bearing surface and 160 mm, blade and security screw.
PTFE, shaft made of DURAN® tubing.
Order Now
SPECIFICATIONS
Environmentally Preferable