Stirrers with stirrer bearing
Supplier: Lenz Laborglas GmbH & CO.KG
06310000 06310503 06310501 06310505 06310502 06310506 06310504 06310507
LENZ06310000EA 0 SGD
LENZ06310000 LENZ06310503 LENZ06310501 LENZ06310505 LENZ06310502 LENZ06310506 LENZ06310504 LENZ06310507
Stirrers with stirrer bearing
Stirrers Stirring Tools
DURAN®, borosilicate glass, PTFE blade and securing screw. Gas tight closure for vacuum stirring apparatus up to 10⁻³ mbar at continuous operational speed of 800 min⁻¹.
  • Ideal for use with 250 to 2000 ml round bottomed flasks
  • Includes PP screw cap with GL 25 closure
Order Now
SPECIFICATIONS
Environmentally Preferable