Prepared microscope slides: typical monocot roots
Supplier: LIEDER JOHANNES
AS214C AS215C AS217C AS222C
LIEDAS214CEA 0 SGD
LIEDAS214C LIEDAS215C LIEDAS217C LIEDAS222C
Prepared microscope slides: typical monocot roots
Slides Prepared Slides
Prepared microscope slides in systematic order showing the anatomy of typical monocot roots in angiospermae.
(ts) total structure