Suspended solids colorimeter, MD100
Supplier: Lovibond
276150
LOVI276150EA 0 SGD
LOVI276150
Suspended solids colorimeter, MD100
Environmental Test Kits
The MD 100 photometer is a portable handheld instrument with a simple button layout, robust waterproof design and large backlit display that are ideal for testing in the field or laboratory conditions.
  • Accurate (3% FS), reproducible results using high quality interference filter with long-life LED
  • Internal ring memory for 16 data sets
  • Real time clock and date
  • OTZ (One Time Zero setting) on versions except those denoted*
  • Optional infrared interface for transfer of test data to PC or printer
Delivery information: Instrument supplied with carry case, 4× AAA batteries, 3× round glass vials with lids, relevant reagents, and instruction manual.