Comparator kits, multiparameter for water testing, Comparator 2000+
Supplier: Lovibond
413550 413570
LOVI413550EA 0 SGD
LOVI413550 LOVI413570
Comparator kits, multiparameter for water testing, Comparator 2000+
Colorimeters Comparator Kits
Lovibond® Comparator system 2000+ is an extremely versatile, modular system for testing water. The battery operated portable lighting unit can be integrated to give standardised lighting conditions to enhance reproducibility.
  • Compact and portable
  • Simple to use yet uncompromising in terms of precision and reproducibility of results
Delivery information: Comparator test kits are supplied as a complete system in a sturdy plastic case. Together with the Comparator 2000+ and test discs, each kit includes all the necessary cells, accessories and Lovibond®More Product Information