L(+)-Potassium sodium tartrate

Supplier: Lovibond

466101
LOVI466101EA 0 SGD
LOVI466101
L(+)-Potassium sodium tartrate
L(+)-Potassium sodium tartrate
Formula: C₄H₄KNaO₆
Melting Pt: 90…100 °C
Density: 1,79 g/cm³ (20 °C)
MDL Number: MFCD00065391
CAS Number: 304-59-6
EINECS: 206-156-8
Merck Index: 12,07840

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR