Phenol USP, Macron Fine Chemicals™

Supplier: Avantor Performance Materials

MACR0610-06EA 641 SGD
MACR0610-06
Phenol USP, Macron Fine Chemicals™
Phenol
Formula: C₆H₅OH
MDL Number: MFCD00002143
CAS Number: 108-95-2
UN: 1671
ADR: 6.1,II

Order Now

Specification Test Results

Meets U.S.P Requirements
GMP Manufactured Product
Meets J.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical = 89.0 %
USP - Assay (C₆H₅OH) ≥89.0 %
USP - Benzene ≤2.0 ppm
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Clarity of Solution and Reaction Passes Test
USP - Nonvolatile Residue ≤0.05 %
Distilling Range - Initial Temperature (For Information Only) = 182.5 °C
Distilling Range - Final Temperature ≤182.5 °C
Stabilizer Passes Test
JP - Identification (1) Passes Test
JP - Identification (2) Passes Test
JP - Boiling Point ≤182 °C
JP - Clarity and Color of Solution and Acidity or Alkalinity Passes Test
JP - Residue on Evaporation ≤0.05 %
JP - Assay ≥88.0 %
Appearance (colorless to slightly pink liquid)
No Class 2 or 3 solvents are expected to be present. The Class 1
solvent Benzene is used as a raw material in a precursor to phenol. The product is tested for any presence of benzene and the result
reported on the Certificate of Analysis.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR