Sodium iodide, granular, AR®, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Warning

Synonyms: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril

MACR1141-12EA 0 SGD
MACR1141-12
Sodium iodide, granular, AR®, Macron Fine Chemicals™
Sodium iodide
Formula: NaI
Boiling Pt: 1300 °C (1013 hPa)
Melting Pt: 662 °C
Density: 3,67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9,III
Merck Index: 13,08705

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use = 0.02 %
Alkalinity (as NaOH) ≤0.02 %
Aluminum (Al) ≤0.0005 %
Barium (Ba) ≤0.002 %
Chloride and Bromide (as Cl) ≤0.01 %
Heavy Metals (as Pb) ≤0.0005 %
Insoluble Matter ≤0.005 %
Iodate (IO₃) ≤0.0003 %
Iron (Fe) ≤5 ppm
Lead (Pb), For Information Only = 0.001 %
Nitrogen Compounds (as N) ≤0.001 %
Phosphate (PO₄) ≤0.001 %
Potassium (K) ≤0.01 %
Sulfate (SO₄) ≤0.005 %
Solubility Passes Test
Hydrazine Monohydrate ≤0.5 ppm
Appearance (coarse, white granules and powder allfiner than 4 mesh)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR