Benzyl alcohol 98.0-100.5% NF, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Warning

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol

MACR1403-12EA 0 SGD
MACR1403-12 MACR1403-19 MACR1403-25 MACR1403-10
Benzyl alcohol 98.0-100.5% NF, Macron Fine Chemicals™
Benzyl alcohol

Caution: This product is not intended for use in preparing parenteral dosage forms.

Formula: C₆H₅CH₂OH
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: –15,3 °C
Density: 1,05 g/cm³ (20 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical 98.0 - 100.5 %
NF - Assay (C₆H₅CH₂OH) 98.0 - 100.5 %
NF - Acidity Passes Test
NF - Clarity of Solution Passes Test
NF - Color of Solution Passes Test
NF - Identification Passes Test
NF - Residue on Evaporation ≤0.05 %
NF - Peroxide Value ≤5
NF - Refractive Index, n²ºD 1.538 - 1.541
NF - Benzaldehyde and Other Related Substances - Benzaldehyde ≤0.15 %
NF - Benzaldehyde and Other Related Substances - Cyclohexylmethanol ≤0.10 %
NF - Benzaldehyde and Other Related Substances - Total Other Peaks Less ≤0.04 %
Than RT of Benzyl Alcohol ≤0.04 %
NF - Benzaldehyde and Other Related Substances - Total Other Peaks ≤0.3 %
Greater Than RT of Benzyl Alcohol ≤0.3 %
NF - Benzene ≤2 ppm
Solubility Passes Test
The Class 1 residual solvent Benzene is likely to be present below 2 ppm. The actual analysis is reported for each lot. The Class 2 solvent Toluene is likely to be present and is process controlled to <30 ppm. No other Class 1, 2 or 3 residual solvents are likely to be present.
This product is not intended for use in preparing parenteral dosage forms.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR