Lactic acid 85%, AR® ACS, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Danger

Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid

Management of Change (MOC) category = N

MACR2676-04EA 0 SGD
MACR2676-04
Lactic acid 85%, AR® ACS, Macron Fine Chemicals™
Lactic acid
Formula: H₃CCH(OH)COOH
MW: 90.08 g/mol
Density: 1.20 g/cm³
Flash Pt: 112.00 °C
MDL Number: MFCD00004520
CAS Number: 50-21-5
UN: 3265
ADR: 8 II
Merck Index: 13,05351

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃CHOHCOOH) 85.0 - 90.0 %
ACS - Chloride (Cl) ≤ 0.001 %
ACS - Heavy Metals (by ICP-OES) ≤ 5 ppm
ACS - Iron (Fe) ≤ 5 ppm
ACS - Residue after Ignition ≤ 0.02 %
ACS - Substances Darkened by H₂SO₄ Passes Test
ACS - Sulfate (SO₄) ≤ 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR